Learn More
Electron Microscopy Sciences EMS-Mount 30 ML
SDP
Catalog No. 5034371
Encompass_Preferred
EMS-Mount is a non-permanent, low viscosity an aqueous mounting medium designed for mounting of tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents. This mounting medium preserves Fast red, Aminoethylycarbazole (AEC), BCIP/NBT, BCIP/INT chromogens and is also compatible with counterstain like Hematoxylin and Nuclear fast red (NFR). It is also suitable for chromogens like DAB and DAB with nickel and cobalt. (It is not compatible with H and E staining).
Application: Mounting of chromogens that cannot be dehydrated with organic solvents. Ready to use. Storage at 2-8 degree C is recommended.
Catalog No. 50-343-71 Supplier Electron Microscopy Sciences Supplier No. 1798505
EMS-Mount is a non-permanent, low viscosity an aqueous mounting medium designed for mounting of tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents. This mounting medium preserves Fast red, Aminoethylycarbazole (AEC), BCIP/NBT, BCIP/INT chromogens and is also compatible with counterstain like Hematoxylin and Nuclear fast red (NFR). It is also suitable for chromogens like DAB and DAB with nickel and cobalt. (It is not compatible with H and E staining).
Application: Mounting of chromogens that cannot be dehydrated with organic solvents. Ready to use. Storage at 2-8 degree C is recommended.