Learn More
Electron Microscopy Sciences EMS Engraving Tool, 7" in Height 1 kit
SDP
Supplier:  Electron Microscopy Sciences 7209601
Encompass_Preferred
Engraver engraves test tubes, bottles, racks, keys, and anything made from glass, metal, stone as well as bone. Engravings are permanent and will not disappear in the presence of chemicals and solvents. The Engraver is 7" in height ( 17.8cm). The Engraver comes with a battery pack and is operated on two AA Batteries.
Catalog No. 50-365-776