Learn More
Electron Microscopy Sciences EMS Coverslip Holder Double
SDP
Catalog No. NC1478807
Encompass_Preferred
EMS introduces an alternative to the standard aluminum slide used to visualize live microscopic organisms (both dorsal and ventral sides) using a pair of coverslips. The holders are made from refined ABS plastic and are reusable. Total slide dimensions: 3" x 1" x 0.045". Well dimensions: 25 mm x 23 mm. Inside opening dimensions: 22 mm x 16 mm. Recommended coverslip: 22 mm x 22 mm Holds coverslip sandwich in place w/out glue. Holds living material between the coverslips, generally in water.
Catalog No. NC1478807 Supplier Electron Microscopy Sciences Supplier No. 7226910
EMS introduces an alternative to the standard aluminum slide used to visualize live microscopic organisms (both dorsal and ventral sides) using a pair of coverslips. The holders are made from refined ABS plastic and are reusable. Total slide dimensions: 3" x 1" x 0.045". Well dimensions: 25 mm x 23 mm. Inside opening dimensions: 22 mm x 16 mm. Recommended coverslip: 22 mm x 22 mm Holds coverslip sandwich in place w/out glue. Holds living material between the coverslips, generally in water.