Learn More
Electron Microscopy Sciences EMS Coverslip Holder Coverslip Holder Single
SDP
Catalog No. NC1473078
Encompass_Preferred
EMS introduces an alternative to the standard aluminum slide used to visualize live microscopic organisms (both dorsal and ventral sides) using a pair of coverslips. The holders are made from refined ABS plastic and are reusable. Total slide dimensions: 3" x 1" x 0.045". Well dimensions: 25 mm x 23 mm. Inside opening dimensions: 22 mm x 16 mm. Recommended coverslip: 22 mm x 22 mm Designed to hold a pair of cover slips. Allows a specimen to be viewed from both sides on a confocal microscope.
Catalog No. NC1473078 Supplier Electron Microscopy Sciences Supplier No. 7226801
EMS introduces an alternative to the standard aluminum slide used to visualize live microscopic organisms (both dorsal and ventral sides) using a pair of coverslips. The holders are made from refined ABS plastic and are reusable. Total slide dimensions: 3" x 1" x 0.045". Well dimensions: 25 mm x 23 mm. Inside opening dimensions: 22 mm x 16 mm. Recommended coverslip: 22 mm x 22 mm Designed to hold a pair of cover slips. Allows a specimen to be viewed from both sides on a confocal microscope.