Learn More
Electron Microscopy Sciences EMS Syntheric Fiber Tweezers Set
SDP
Catalog No. 5023983
Encompass_Preferred
Continuous use temp to 150 degree C. High purity, chemical resistant, ESD safe. Clean room and medical devices approved with low extraction value. Made from SV plastic based on polyvinylidene fluoride (PVDF) for good mechanical strength and toughness, smooth surface. Excellent chemical resistance to most aggressive substances. Set of five tweezers styles 702A, 705, 707, 708, 709, 710.
Catalog No. 50-239-83 Supplier Electron Microscopy Sciences Supplier No. 7846006
Continuous use temp to 150 degree C. High purity, chemical resistant, ESD safe. Clean room and medical devices approved with low extraction value. Made from SV plastic based on polyvinylidene fluoride (PVDF) for good mechanical strength and toughness, smooth surface. Excellent chemical resistance to most aggressive substances. Set of five tweezers styles 702A, 705, 707, 708, 709, 710.