Learn More
Electron Microscopy Sciences EMS Syntheric Fiber Tweezers Style 710, 116 mm OAL
SDP
Catalog No. 5023985
Encompass_Preferred
Continuous use temp to 150 degree C. High purity, chemical resistant, ESD safe. Clean room and medical devices approved with low extraction value. Made from SV plastic based on polyvinylidene fluoride (PVDF) for good mechanical strength and toughness, smooth surface. Excellent chemical resistance to most aggressive substances.
OAL: 116 mm
Catalog No. 50-239-85 Supplier Electron Microscopy Sciences Supplier No. 78460710
Continuous use temp to 150 degree C. High purity, chemical resistant, ESD safe. Clean room and medical devices approved with low extraction value. Made from SV plastic based on polyvinylidene fluoride (PVDF) for good mechanical strength and toughness, smooth surface. Excellent chemical resistance to most aggressive substances.
OAL: 116 mm