Learn More
Electron Microscopy Sciences EMS Film Racks for 3 1/4 x4" Film
SDP
Catalog No. 5025781
Encompass_Preferred
Fabricated acrylic racks that are designed to fit with Kodak developing tanks. Sheet film is firmly held in the slots in the side rails while solution is freely circulating inside the tank. Measurements 3.5 x 6-7/8 x 6"H. Holds 18 sheets of film 3 1/4 x 4"
Catalog No. 50-257-81 Supplier Electron Microscopy Sciences Supplier No. 74350
Fabricated acrylic racks that are designed to fit with Kodak developing tanks. Sheet film is firmly held in the slots in the side rails while solution is freely circulating inside the tank. Measurements 3.5 x 6-7/8 x 6"H. Holds 18 sheets of film 3 1/4 x 4"