Learn More
Electron Microscopy Sciences EMS Germinator 500™ Germinator 500 Dry Sterilizer 110 Volt
SDP
Catalog No. 50367182
Encompass_Preferred
The GERMINATOR 500 is a simple, inexpensive way to quickly decontaminate your micro-dissecting instruments between procedures. The unique stainless steel glass bead bath remains at a constant 500 degrees F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them, usually taking only 15 seconds.
The operating temperature for the unit is 500°F 15° (233°C). No messy chemicals and no need for open flames in the lab.
The unit measures 6.75'' x 5.25'' x 5.25'' (H x W x D) and the Stainless Steel Well measures: 2.0'' diameter x 4.0'' deep(ID). The unit only weighs 4.5lbs.
Catalog No. 50-367-182 Supplier Electron Microscopy Sciences Supplier No. 6611810
The GERMINATOR 500 is a simple, inexpensive way to quickly decontaminate your micro-dissecting instruments between procedures. The unique stainless steel glass bead bath remains at a constant 500 degrees F, allowing you to insert the tips of your instruments whenever you wish to decontaminate them, usually taking only 15 seconds.
The operating temperature for the unit is 500°F 15° (233°C). No messy chemicals and no need for open flames in the lab.
The unit measures 6.75'' x 5.25'' x 5.25'' (H x W x D) and the Stainless Steel Well measures: 2.0'' diameter x 4.0'' deep(ID). The unit only weighs 4.5lbs.