Learn More
Electron Microscopy Sciences EMS Glyoxal 3% Fixative 100 ML
SDP
Catalog No. 501931591
Encompass_Preferred
Glyoxal 3% Fixative is an alternative to Formalin and Formaldehyde in many different applications. Our unique kit comes premixed and measured and ready to use. The kit consists of 2 vials: Solution A & Solution B (Absolute Ethanol, Glyoxal Solution, Acetic Acid and water).
Catalog No. 50-193-1591 Supplier Electron Microscopy Sciences Supplier No. 16525
Glyoxal 3% Fixative is an alternative to Formalin and Formaldehyde in many different applications. Our unique kit comes premixed and measured and ready to use. The kit consists of 2 vials: Solution A & Solution B (Absolute Ethanol, Glyoxal Solution, Acetic Acid and water).