Learn More
Electron Microscopy Sciences EMS Golden Nylon Brush ½"
SDP
Catalog No. 501931664
Encompass_Preferred
These brushes are perfect for cryomicrotomy – for controlled manipulation of frozen sections in the cryostat, as well as cryomicrotome. Easier to clean than animal hair brushes and more durable.
Catalog No. 50-193-1664 Supplier Electron Microscopy Sciences Supplier No. 6611002
These brushes are perfect for cryomicrotomy – for controlled manipulation of frozen sections in the cryostat, as well as cryomicrotome. Easier to clean than animal hair brushes and more durable.