Learn More
Electron Microscopy Sciences EMS Hobby Time Blades
SDP
Supplier:  Electron Microscopy Sciences 71982CS
Encompass_Preferred
Hobby blades are used in a wide range of applications due to the simple but precise cutting tip. These 0.021" carbon steel blades fit most standard handles. They are precision ground for sharpness and durability. The Handles are made from aluminum construction for durability and they have a 4 jaw check which holds the blade securely.
Catalog No. 50-192-7061