Learn More
Electron Microscopy Sciences EMS Iris Scissors Premium with Angled Blades
SDP
Catalog No. NC1588309
Encompass_Preferred
These Iris scissors are made with sharp tips and are perfect for precise vascular and neurological dissection; ideal for small specimens and animal dissection.
OAL: 114 mm
Catalog No. NC1588309 Supplier Electron Microscopy Sciences Supplier No. 7299818
These Iris scissors are made with sharp tips and are perfect for precise vascular and neurological dissection; ideal for small specimens and animal dissection.
OAL: 114 mm