Learn More
Grainger Fire-Dex EMS Jacket, Unisex, XL, Regular, Navy, Aramid Fiber/PTFE
Catalog No. 502773139
Encompass_Preferred
This unisex EMS jacket is engineered for emergency medical professionals to provide essential protection against line-of-duty hazards. Constructed with a moisture barrier, the jacket is both waterproof and bloodborne pathogen-resistant for enhanced safety in diverse environments.
  • Features an EMS moisture barrier
  • Constructed from aramid fiber and PTFE materials
  • Includes reflective triple trim
  • Equipped with two semi-bellows hand-warmer pockets, a radio pocket, and a patch pocket
  • Includes two mic clips on the chest
  • Designed with woven hook and loop cuff adjusters
  • Features an inner zipper and outer hook and loop closure
  • Compatible with optional zip-in fleece or nylon liners
  • Meets NFPA 1950 (1999) standards
Catalog No. 50-277-3139 Supplier Grainger Supplier No. 13T331
This unisex EMS jacket is engineered for emergency medical professionals to provide essential protection against line-of-duty hazards. Constructed with a moisture barrier, the jacket is both waterproof and bloodborne pathogen-resistant for enhanced safety in diverse environments.
  • Features an EMS moisture barrier
  • Constructed from aramid fiber and PTFE materials
  • Includes reflective triple trim
  • Equipped with two semi-bellows hand-warmer pockets, a radio pocket, and a patch pocket
  • Includes two mic clips on the chest
  • Designed with woven hook and loop cuff adjusters
  • Features an inner zipper and outer hook and loop closure
  • Compatible with optional zip-in fleece or nylon liners
  • Meets NFPA 1950 (1999) standards