Learn More
Electron Microscopy Sciences EMS Magnifier Forceps, 5½"
SDP
Catalog No. 501931458
Encompass_Preferred
These forceps feature extra-fine serrated tips with a ribbed grip very helpful for tight extractions. The magnifier provides 10x magnification to the extraction field. Made of stainless steel.
OAL: 5½"
Catalog No. 50-193-1458 Supplier Electron Microscopy Sciences Supplier No. 7299745
These forceps feature extra-fine serrated tips with a ribbed grip very helpful for tight extractions. The magnifier provides 10x magnification to the extraction field. Made of stainless steel.
OAL: 5½"