Learn More
Electron Microscopy Sciences EMS Microscope Slide Holder, 3.75" W x 15.25" L (95 mm x 387 mm)
SDP
Catalog No. 501931275
Encompass_Preferred
A universal slide holder that is ideal for drying slides (or drying and organizing slides) during frozen sectioning or routine processing. Made of durable polyethylene, with 12 slide slots designed to hold slides vertically or horizontally. Dimensions: 3.75" W x 15.25" L (95 mm x 387 mm).
Dimensions: 3.75" W x 15.25" L (95 mm x 387 mm)
Catalog No. 50-193-1275 Supplier Electron Microscopy Sciences Supplier No. 7222008
A universal slide holder that is ideal for drying slides (or drying and organizing slides) during frozen sectioning or routine processing. Made of durable polyethylene, with 12 slide slots designed to hold slides vertically or horizontally. Dimensions: 3.75" W x 15.25" L (95 mm x 387 mm).
Dimensions: 3.75" W x 15.25" L (95 mm x 387 mm)