Learn More
Electron Microscopy Sciences Micro-Slide Cabinet
SDP
Catalog No. 5028758
Encompass
Our newly introduced cabinets for filing 3x1" slides is the most effective system available to the ever-growing problem of slide storage and retrieval. They are compact, sturdy and economical. The cabinets are stackable and are secured by interlocks. Store up to 54,000 slides in only 2 square feet of space. Low cost slide storage. Ideal for use in Pathology, Cytology, Hematology, Histology, Dermatology, Research, Education. Stackable cabinets for 3x1" slides. 750 slides per drawer. 6 drawers.
This item is not returnable. View return policy
Catalog No. 50-287-58 Supplier Electron Microscopy Sciences Supplier No. 7478020
Our newly introduced cabinets for filing 3x1" slides is the most effective system available to the ever-growing problem of slide storage and retrieval. They are compact, sturdy and economical. The cabinets are stackable and are secured by interlocks. Store up to 54,000 slides in only 2 square feet of space. Low cost slide storage. Ideal for use in Pathology, Cytology, Hematology, Histology, Dermatology, Research, Education. Stackable cabinets for 3x1" slides. 750 slides per drawer. 6 drawers.