Learn More
Electron Microscopy Sciences EMS Molded Flat Embedding Mold
SDP
Catalog No. 50930539
Encompass_Preferred
Molds are injected molded polyethylene that is resistant to most embedding materials and will easily peeled-off the blocks. Mold fits perfectly inside the cocoon box BE8 (64300-8). These molds are reusable. Each cavity measures 12mm(L) x 5mm(W) x 3.5mm Deep. Overall mold measures: 94 x 56 x 14mm.
Catalog No. 50-930-539 Supplier Electron Microscopy Sciences Supplier No. 7090512
Molds are injected molded polyethylene that is resistant to most embedding materials and will easily peeled-off the blocks. Mold fits perfectly inside the cocoon box BE8 (64300-8). These molds are reusable. Each cavity measures 12mm(L) x 5mm(W) x 3.5mm Deep. Overall mold measures: 94 x 56 x 14mm.