Learn More
Electron Microscopy Sciences EMS Rapid Immersion Freezer Muscle Biopsy Clamps
SDP
Catalog No. 50980724
Encompass_Preferred
Clamps designed for use with the EMS-002 Rapid Immersion Freezer for human muscle biopsies to keep the muscle from contracting during fixation. Use improves the morphology of the sample by rapid freezing by plunging into liquid cryogen.
Catalog No. 50-980-724 Supplier Electron Microscopy Sciences Supplier No. 37459
Clamps designed for use with the EMS-002 Rapid Immersion Freezer for human muscle biopsies to keep the muscle from contracting during fixation. Use improves the morphology of the sample by rapid freezing by plunging into liquid cryogen.