Learn More
Electron Microscopy Sciences EMS PHEM Buffer 500 ML
SDP
Catalog No. 501931295
Encompass_Preferred
PHEM buffer has been used primarily for tissues and cell cultures being processed for immunocytochemical studies. Most antigens, especially intra cellular ones, stain better using PHEM than those processed using PBS, it also has a more limited effect on biochemical reactions and enzymes. Very good preservation of cellular ultrastructure, notably microtubules, is also an advantage of PHEM use.
Catalog No. 50-193-1295 Supplier Electron Microscopy Sciences Supplier No. 11163
PHEM buffer has been used primarily for tissues and cell cultures being processed for immunocytochemical studies. Most antigens, especially intra cellular ones, stain better using PHEM than those processed using PBS, it also has a more limited effect on biochemical reactions and enzymes. Very good preservation of cellular ultrastructure, notably microtubules, is also an advantage of PHEM use.