Learn More
Electron Microscopy Sciences EMS EMS Poly III Evaporation-Controlled Automated Embedding and Polymerization
SDP
Catalog No. 501927565
Encompass_Preferred
The EMS Poly III reduces lengthy and sometimes tedious manual embedding procedures for electron microscopy to a few simple steps for up to 52 vials. It uses a proper combination of pressure and temperature. The specimen chamber is temperature controlled from room temperature to 70 degree C. The pressure in the chamber can be reduced from ambient pressure with an inbuilt vacuum-pump that controls pressure to within narrow limits and warrants solvents will evaporate without the risk of explosive boiling. The exhaust of the vacuum pump can vent into a fume hood.
The instrument modifiable preset programs are optimized for an efficient removal of solvent from specimens eliminating the risk of boiling. Bulk removal of solvent is followed by the thorough removal of trace amounts.
Load at the end of a workday and (when using acetone or propylene oxide) the next morning are ready for polymerization after being topped off with pure resin.
Catalog No. 50-192-7565 Supplier Electron Microscopy Sciences Supplier No. 4444
Add to Cart
Add to Cart
Request a Quote
The EMS Poly III reduces lengthy and sometimes tedious manual embedding procedures for electron microscopy to a few simple steps for up to 52 vials. It uses a proper combination of pressure and temperature. The specimen chamber is temperature controlled from room temperature to 70 degree C. The pressure in the chamber can be reduced from ambient pressure with an inbuilt vacuum-pump that controls pressure to within narrow limits and warrants solvents will evaporate without the risk of explosive boiling. The exhaust of the vacuum pump can vent into a fume hood.
The instrument modifiable preset programs are optimized for an efficient removal of solvent from specimens eliminating the risk of boiling. Bulk removal of solvent is followed by the thorough removal of trace amounts.
Load at the end of a workday and (when using acetone or propylene oxide) the next morning are ready for polymerization after being topped off with pure resin.