Learn More
Electron Microscopy Sciences EMS ProCut Scissors are designed for users who need a high-performing, long-lasting, and cost-effective solution for a wide range of applications, including both industrial and life science settings.
Supplier: Electron Microscopy Sciences 362S35.NP.EM
The EMS ProCut line is conceived for those professional users that need high-performing and long-lasting products with a cost-effective solution for a wide range of applications. This line includes the most common geometries and iconic design. Made of fine steel, hardened and tempered with state-of-the art technologies, hand finished by artisans and double nickel-plated to provide a shield against wear and corrosion, these scissors have all the features needed for industrial use and general applications.