Learn More
Electron Microscopy Sciences EMS ProCut Scissors are designed for users who need a high-performing, long-lasting, and cost-effective solution for a wide range of applications, including both industrial and life science settings.
SDP
Catalog No. NC2828552
Encompass_Preferred
The EMS ProCut line is conceived for those professional users that need high-performing and long-lasting products with a cost-effective solution for a wide range of applications. This line includes the most common geometries and iconic design. Made of fine steel, hardened and tempered with state-of-the art technologies, hand finished by artisans and double nickel-plated to provide a shield against wear and corrosion, these scissors have all the features needed for industrial use and general applications.
Catalog No. NC2828552 Supplier Electron Microscopy Sciences Supplier No. 372SY50.NP.EM
The EMS ProCut line is conceived for those professional users that need high-performing and long-lasting products with a cost-effective solution for a wide range of applications. This line includes the most common geometries and iconic design. Made of fine steel, hardened and tempered with state-of-the art technologies, hand finished by artisans and double nickel-plated to provide a shield against wear and corrosion, these scissors have all the features needed for industrial use and general applications.