Learn More
Electron Microscopy Sciences EMS Rapid Immersion Freezer Propane Transfer System
SDP
Catalog No. 50980718
Encompass_Preferred
Propane transfer system for use with the EMS-002 Rapid Immersion Freezer allows for the efficient transfer of propane to the condensation chamber fromthe propane storage vessel.
Catalog No. 50-980-718 Supplier Electron Microscopy Sciences Supplier No. 37015
Propane transfer system for use with the EMS-002 Rapid Immersion Freezer allows for the efficient transfer of propane to the condensation chamber fromthe propane storage vessel.