Learn More
Electron Microscopy Sciences EMS Rubber Tipped Forceps 8"
SDP
Catalog No. 501931265
Encompass_Preferred
Soft rubber rounded tipped forceps allow for secure dissection with minimal tissue damage or compression. An excellent option for techniques that require no metal exposure. Blue tips provide excellent contrast for specimen photography. Warning: These forceps are not autoclavable. To clean rince under water or wipe clean with any laboratory disinfectant. Made of standard grade stainless steel.
OAL: 8"
Catalog No. 50-193-1265 Supplier Electron Microscopy Sciences Supplier No. 7299704
Soft rubber rounded tipped forceps allow for secure dissection with minimal tissue damage or compression. An excellent option for techniques that require no metal exposure. Blue tips provide excellent contrast for specimen photography. Warning: These forceps are not autoclavable. To clean rince under water or wipe clean with any laboratory disinfectant. Made of standard grade stainless steel.
OAL: 8"