Learn More
Electron Microscopy Sciences EMS Shield Mount with Dabco 30 ML
SDP
Catalog No. 5034375
Encompass_Preferred
EMS Shield Mount  is an aqueous non-permanent, low viscosity mounting medium for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes.
Application: Use for immunofluorescence and confocal microscopy.
Reagent: Ready to use mounting medium for Immunofluorescence.
Catalog No. 50-343-75 Supplier Electron Microscopy Sciences Supplier No. 17985150
EMS Shield Mount  is an aqueous non-permanent, low viscosity mounting medium for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes.
Application: Use for immunofluorescence and confocal microscopy.
Reagent: Ready to use mounting medium for Immunofluorescence.