Learn More
Electron Microscopy Sciences EMS Slide Staining Tray Kit
SDP
Catalog No. 501931269
Encompass_Preferred
With a removable pan and rack, this slide staining tray kit can be used in various areas of the lab including microbiology, gram staining in histology, and special staining in hematology. Full kit includes base with attached leveling indicator, four leveling feet, removable rack, and removable tray. Made of stainless steel.
Catalog No. 50-193-1269 Supplier Electron Microscopy Sciences Supplier No. 7222001
With a removable pan and rack, this slide staining tray kit can be used in various areas of the lab including microbiology, gram staining in histology, and special staining in hematology. Full kit includes base with attached leveling indicator, four leveling feet, removable rack, and removable tray. Made of stainless steel.