Learn More
Electron Microscopy Sciences EMS Specimen Paddle Forceps, 1 ea
SDP
Catalog No. 501931262
Encompass_Preferred
Paddle forceps hold all types of tissue with the secure ribbed grip needed to accomplish thin sections for histology analysis. Thin sections are essential for proper infiltration of reagents prior to histology study. Great for training residents and technicians on thin section technique, cutting thin sections of fatty tissue like breast tissue and cutting even thickness sections of any type tissue. Made of standard grade stainless steel.
Catalog No. 50-193-1262 Supplier Electron Microscopy Sciences Supplier No. 7299701
Paddle forceps hold all types of tissue with the secure ribbed grip needed to accomplish thin sections for histology analysis. Thin sections are essential for proper infiltration of reagents prior to histology study. Great for training residents and technicians on thin section technique, cutting thin sections of fatty tissue like breast tissue and cutting even thickness sections of any type tissue. Made of standard grade stainless steel.