Learn More
Electron Microscopy Sciences Sure-Cut Surfactant 50 GR
SDP
Catalog No. 5029166
Encompass_Preferred
Epoxy resins are easier to section with glass, as well as diamond knives, when the embedding medium contains 0.5-1.0% of our SURE-CUT. The life of the diamond knife will be extended with its use because it will reduce the friction of the plastic block during sectioning.
Catalog No. 50-291-66 Supplier Electron Microscopy Sciences Supplier No. 21630
Epoxy resins are easier to section with glass, as well as diamond knives, when the embedding medium contains 0.5-1.0% of our SURE-CUT. The life of the diamond knife will be extended with its use because it will reduce the friction of the plastic block during sectioning.