Learn More
Electron Microscopy Sciences EMS Tissue Capture Pen, Wide Point
SDP
Catalog No. 50930983
Encompass_Preferred
With one swift application of the EMS Tissue Capture Pen, you are able to change your plain glass slide into a coated one. The EMS Tissue Capture Pen is a coating pen with a wide point (18 mm) for fixing various tissue sections (paraffin embedded, frozen and formalin sections, etc). The unique pen is unaffected by heat up to 120 degree C so it offers secure and firm adhesion of the tissue section.
One application is all that is needed, and after it is applied, allow to dry for 1-2 minutes.
The EMS Tissue Capture may be used in conjunction with our Pap-Pen and will allow for easy placement of adherent as well as nonadherent cells on a glass slide. Immunostaining/ISH can be performed by applying a few drops of tissue suspension or liquid media onto the glass slide which is circled by the Pap-Pen, and then your section can be placed. Can be used in microwave staining, immunostaining, in situ, etc.
Catalog No. 50-930-983 Supplier Electron Microscopy Sciences Supplier No. 7131410
With one swift application of the EMS Tissue Capture Pen, you are able to change your plain glass slide into a coated one. The EMS Tissue Capture Pen is a coating pen with a wide point (18 mm) for fixing various tissue sections (paraffin embedded, frozen and formalin sections, etc). The unique pen is unaffected by heat up to 120 degree C so it offers secure and firm adhesion of the tissue section.
One application is all that is needed, and after it is applied, allow to dry for 1-2 minutes.
The EMS Tissue Capture may be used in conjunction with our Pap-Pen and will allow for easy placement of adherent as well as nonadherent cells on a glass slide. Immunostaining/ISH can be performed by applying a few drops of tissue suspension or liquid media onto the glass slide which is circled by the Pap-Pen, and then your section can be placed. Can be used in microwave staining, immunostaining, in situ, etc.