Learn More
Electron Microscopy Sciences EMS Tweezers Style 000, SA steel, Flat edges, strong with radiused edges Tips
SDP
Catalog No. 5023869
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Flat edges, strong with radiused edges
OAL: 120 mm
Catalog No. 50-238-69 Supplier Electron Microscopy Sciences Supplier No. 78312000
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Flat edges, strong with radiused edges
OAL: 120 mm