Learn More
Electron Microscopy Sciences EMS Tweezers Style 00B, SA steel, Flat edges, strong Tips
SDP
Catalog No. 5023870
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Serrated grips.
Material: SA Steel
Tips: Flat edges, strong
OAL: 120 mm
Catalog No. 50-238-70 Supplier Electron Microscopy Sciences Supplier No. 7831300B
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Serrated grips.
Material: SA Steel
Tips: Flat edges, strong
OAL: 120 mm