Learn More
Electron Microscopy Sciences EMS Tweezers Style 00D, SA steel, Flat edges, strong with serration Tips
SDP
Catalog No. 5023871
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Serrated grips.
Material: SA Steel
Tips: Flat edges, strong with serration
OAL: 120 mm
Catalog No. 50-238-71 Supplier Electron Microscopy Sciences Supplier No. 7831400D
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Serrated grips.
Material: SA Steel
Tips: Flat edges, strong with serration
OAL: 120 mm