Learn More
Electron Microscopy Sciences EMS Tweezers Style 1, SA steel, Fine, accurate, strong Tips
SDP
Catalog No. 5023853
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine, accurate, strong
OAL: 120 mm
Catalog No. 50-238-53 Supplier Electron Microscopy Sciences Supplier No. 783161
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine, accurate, strong
OAL: 120 mm