Learn More
Electron Microscopy Sciences EMS Tweezers Style 10G, SA steel, Fine Tips
SDP
Catalog No. 5023887
Encompass_Preferred
The tweezers are the Swiss highest quality. General purpose use tweezers. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine
OAL: 110 mm
Catalog No. 50-238-87 Supplier Electron Microscopy Sciences Supplier No. 7832410G
The tweezers are the Swiss highest quality. General purpose use tweezers. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Fine
OAL: 110 mm