Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Tweezers Style 120A, 110 mm
SDP

Supplier:  Electron Microscopy Sciences 78355120A

Encompass_Preferred

Very well made tweezers that serves a wide range of applications, from electronics to dentistry and anatomy. Extremely accurate serration quality.
OAL: 110 mm
Tips: Short fine and sturdy tips

Catalog No. 50-306-19


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.