Learn More
Electron Microscopy Sciences EMS Flat-Tip Tweezers Style 125A, 120 mm OAL
SDP
Catalog No. 5023959
Encompass_Preferred
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Smooth, flat, and elongated, no scratch tips.. Tip: 2.7mm wide.
OAL: 120 mm
Catalog No. 50-239-59 Supplier Electron Microscopy Sciences Supplier No. 78332125A
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Smooth, flat, and elongated, no scratch tips.. Tip: 2.7mm wide.
OAL: 120 mm