Learn More
Electron Microscopy Sciences EMS Tweezers Style 15ASW, Carbon Steel, 10 mm curved cutting jaws Tips
SDP
Catalog No. 5023903
Encompass_Preferred
The tweezers are the Swiss highest quality.
Material: Carbon Steel
Tips: 10 mm curved cutting jaws
OAL: 115 mm
Catalog No. 50-239-03 Supplier Electron Microscopy Sciences Supplier No. 7836015ASW
The tweezers are the Swiss highest quality.
Material: Carbon Steel
Tips: 10 mm curved cutting jaws
OAL: 115 mm