Learn More
Electron Microscopy Sciences EMS Tweezers Style 2, SA steel, Strong, sharp Tips
SDP
Catalog No. 5023855
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Strong, sharp
OAL: 120 mm
Catalog No. 50-238-55 Supplier Electron Microscopy Sciences Supplier No. 783172
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Strong, sharp
OAL: 120 mm