Learn More
Electron Microscopy Sciences EMS Tweezers Style 2A, Titanium alloy, Straight Tips
SDP
Catalog No. 5023845
Encompass_Preferred
The tweezers are the Swiss highest quality. Titanium alloy is low density and has good mechanical properties and high temp resistance (1600 deg F). Non magnetic. Good corrosion resistance. Large flat gripping surface for handling without puncturing.
Material: Titanium alloy
Tips: Straight
OAL: 120 mm
Catalog No. 50-238-45 Supplier Electron Microscopy Sciences Supplier No. 729192ATA
The tweezers are the Swiss highest quality. Titanium alloy is low density and has good mechanical properties and high temp resistance (1600 deg F). Non magnetic. Good corrosion resistance. Large flat gripping surface for handling without puncturing.
Material: Titanium alloy
Tips: Straight
OAL: 120 mm