Learn More
Electron Microscopy Sciences EMS Tweezers Style 2A, PTFE Coating, Straight Tips
SDP
Catalog No. 5023874
Encompass_Preferred
The tweezers are the Swiss highest quality. Provides a larger flat gripping surface for better handling of small objects without puncturing. PTFE is a superior anti-scratch, non-stick material.
Material: PTFE Coating
Tips: Straight
OAL: 120 mm
Catalog No. 50-238-74 Supplier Electron Microscopy Sciences Supplier No. 729192ATE
The tweezers are the Swiss highest quality. Provides a larger flat gripping surface for better handling of small objects without puncturing. PTFE is a superior anti-scratch, non-stick material.
Material: PTFE Coating
Tips: Straight
OAL: 120 mm