Learn More
Electron Microscopy Sciences EMS Tweezers Style 2AB, SA steel, Curved tips Tips
SDP
Catalog No. 5023846
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel is used where corrosion resistance and toughness are primary requirements. Anti-glare satin finish. Provides a larger flat gripping surface for handling of small objects without puncturing.
Material: SA Steel
Tips: Curved tips
OAL: 120 mm
Catalog No. 50-238-46 Supplier Electron Microscopy Sciences Supplier No. 729192AB
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel is used where corrosion resistance and toughness are primary requirements. Anti-glare satin finish. Provides a larger flat gripping surface for handling of small objects without puncturing.
Material: SA Steel
Tips: Curved tips
OAL: 120 mm