Learn More
Electron Microscopy Sciences EMS Tweezers Style 2AZ, SA steel, Same tips as 2A but ‘Z’ shaped Tips
SDP
Catalog No. 5023850
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Same tips as 2A but ‘Z’ shaped
OAL: 120 mm
Catalog No. 50-238-50 Supplier Electron Microscopy Sciences Supplier No. 783172AZ
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Same tips as 2A but ‘Z’ shaped
OAL: 120 mm