Learn More
Electron Microscopy Sciences EMS Tweezers Style 3, NC Ultra-Fine, Very fine with superior finish Tips
SDP
Catalog No. 5023873
Encompass_Preferred
The tweezers are the Swiss highest quality.
Grade: NC Ultra-Fine
Tips: Very fine with superior finish
OAL: 120 mm
Catalog No. 50-238-73 Supplier Electron Microscopy Sciences Supplier No. 785183
The tweezers are the Swiss highest quality.
Grade: NC Ultra-Fine
Tips: Very fine with superior finish
OAL: 120 mm