Learn More
Electron Microscopy Sciences EMS Tweezers Style 3, SA steel, Very fine with superior finish Tips
SDP
Catalog No. 5023849
Encompass_Preferred
The tweezers are the Swiss highest quality. Recommended for handling of EM grids. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very fine with superior finish
OAL: 120 mm
Catalog No. 50-238-49 Supplier Electron Microscopy Sciences Supplier No. 783183
The tweezers are the Swiss highest quality. Recommended for handling of EM grids. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
Material: SA Steel
Tips: Very fine with superior finish
OAL: 120 mm