Learn More
Electron Microscopy Sciences EMS Tweezers Style 3R, SA steel, Extra fine Tips
SDP
Catalog No. 5023854
Encompass_Preferred
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Very sharp, reverse action tweezers.
Material: SA Steel
Tips: Extra fine
OAL: 120 mm
Catalog No. 50-238-54 Supplier Electron Microscopy Sciences Supplier No. 783183R
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Very sharp, reverse action tweezers.
Material: SA Steel
Tips: Extra fine
OAL: 120 mm