Learn More
Electron Microscopy Sciences EMS ESD Tweezers Style 3 SA/Dr, SA Steel, 120 mm OAL
SDP
Catalog No. 5023979
Encompass_Preferred
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
OAL: 120 mm
Tips: SA Steel
Material: Very sharp and superior finish
Catalog No. 50-239-79 Supplier Electron Microscopy Sciences Supplier No. 783933
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish.
OAL: 120 mm
Tips: SA Steel
Material: Very sharp and superior finish