Learn More
Electron Microscopy Sciences EMS Flat-Tip Tweezers Style 33A, 115 mm OAL
SDP
Catalog No. 5023966
Encompass_Preferred
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Flat, round tips.. Tip 6.6mm wide.
OAL: 115 mm
Catalog No. 50-239-66 Supplier Electron Microscopy Sciences Supplier No. 7833333A
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Flat, round tips.. Tip 6.6mm wide.
OAL: 115 mm