Learn More
Electron Microscopy Sciences EMS Flat-Tip Tweezers Style 35B, 118 mm OAL
SDP
Catalog No. 5023961
Encompass_Preferred
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Smooth and short curved wide tips.
OAL: 118 mm
Catalog No. 50-239-61 Supplier Electron Microscopy Sciences Supplier No. 7821106
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Smooth and short curved wide tips.
OAL: 118 mm