Learn More
Electron Microscopy Sciences EMS Wafer Tweezers Style 35S
SDP
Catalog No. 5024002
Encompass_Preferred
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 29.0 mm
Depth: 5.5 mm
Offset: 3.0 mm
Catalog No. 50-240-02 Supplier Electron Microscopy Sciences Supplier No. 7841135S
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel with a very smooth surface and a non-glare finish.
Tip Width: 29.0 mm
Depth: 5.5 mm
Offset: 3.0 mm