Learn More
Electron Microscopy Sciences EMS Flat-Tip Tweezers Style 36A, 120 mm OAL
SDP
Catalog No. 5023969
Encompass_Preferred
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Smooth and upward bent wide tips, good for handling delicate parts. Tip 6.2mm wide.
OAL: 120 mm
Catalog No. 50-239-69 Supplier Electron Microscopy Sciences Supplier No. 7833636A
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Smooth and upward bent wide tips, good for handling delicate parts. Tip 6.2mm wide.
OAL: 120 mm